Citric acid monohydrate ≥99% (by acidimetry), granules, BAKER ANALYZED® A.C.S. Reagent ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

Management of Change (MOC) category = N

BAKR0110-07 BAKR0110-05 0110-01
CAJT0110-7EA 2404.49 CAD
CAJT0110-7 CAJT0110-1 CAJT0110-5 JT0110-1
Citric acid monohydrate ≥99% (by acidimetry), granules, BAKER ANALYZED® A.C.S. Reagent ACS, J.T.Baker®
Citric acid monohydrate

The quality of many BAKER ANALYZED™ Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are essential for any chemical stockroom.

Formula: C₆H₈O₇.H₂O
Melting Pt: 135…152 °C
Density: 1.54 g/cm³
Flash Pt: 173.9 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00149972
CAS Number: 5949-29-1
EINECS: 201-069-1
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (HOC(COOH)(CH₂COOH)₂·H₂O) (by acidimetry) 99.0 - 102.0 %
Insoluble Matter ≤0.005 %
Residue on Ignition ≤0.02 %
Chloride (Cl) ≤0.001 %
Oxalate (C₂O₄) Passes Test
Phosphate (PO₄) ≤0.001 %
Sulfur Compounds (as SO₄) ≤0.002 %
Substances Carbonizable by Hot H₂SO₄ Passes Test
Trace Impurities - Iron (Fe) ≤2.0 ppm
Trace Impurities - Lead (Pb) ≤2.000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR