Orthoboric acid ≥99.5% (by acidimetry), granular, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid

Management of Change (MOC) category = N

BAKR0084-07 BAKR0084-08 BAKR0084-19 BAKR0084-05
CAJT0084-7EA 1828.2 CAD
CAJT0084-7 CAJT0084-8 CAJT0084-1 CAJT0084-19 CAJT0084-5
Orthoboric acid ≥99.5% (by acidimetry), granular, BAKER ANALYZED® ACS, J.T.Baker®
ortho-Boric acid

Granular. Lot analysis on label.

Formula: B(OH)₃
MW: 61.83 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.44 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (H₃BO₃) (by acidimetry) ≥99.5 %
Insoluble in Methanol ≤0.005 %
Nonvolatile with Methanol ≤0.05 %
Chloride (Cl) ≤0.001 %
Phosphate (PO₄) ≤0.001 %
Sulfate (SO₄) ≤0.005 %
Calcium (Ca) ≤0.005 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR