Phage Display Competent Cells, Biosearch Technologies
Supplier: Lucigen
60502-1 60502-2 60522-2 60512-1 60512-2 60522-1
95040-450EA 756.54 CAD
95040-450 95040-452 95057-476 95057-470 95057-472 CA95057-474
Phage Display Competent Cells, Biosearch Technologies
Competent Cells
Lucigen’s Phage Display Electrocompetent Cells are perfect for antibody phage display or peptide phage display library creation
Lucigen offers the following Phage Display Electrocompetent Cells:
TG1 Electrocompetent Cells: An amber suppressor strain (supE) prepared as highly efficient (≥4 × 10^10 cfu/µg) electrocompetent cells for phage display library screening. These cells are used for phage display and protein expression.
SS320 Electrocompetent Cells: A non-amber suppressor strain (sometimes called MC1061F') prepared as highly efficient (≥4 × 10^10 cfu/µg) electrocompetent cellsMore Product Information