Corning® TSB (Tryptic Soy Broth), Animal-Free, Corning
Supplier: Corning
46-060-CI 61-411-RO 46-060-CM 46-060-LB
CA45001-156EA 32.24 CAD
CA45001-156 CA45001-226 10810-060 CA71002-820
Corning® TSB (Tryptic Soy Broth), Animal-Free, Corning
Media for Microbiology Dehydrated Media for Microbiology
This Tryptic Soy Broth (TSB) product is offered for use in sterility testing and validation in the manufacture of a liquid product.
Sterile, ready-to-use solution for the imulaiton of aseptic filling of liquids and sterility testing. Due to the rich nutrient base, TSB may be used in the cultivation of fastidious microorganisms. This growth medium is free of animal-sourced components, making it ideal for testing the sterility of animal-free biopharmaceutical processes.
The use of this medium in place of the product intended for production allows an accurate assessment of sterility throughout the manufacture process.
Certifications: Complies with current USP requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable
Shipping temperature Ambient Ambient Ambient
Storage temperature Ambient Ambient Ambient