Corning® Tris Base Buffer, Powder, Corning
Supplier: Corning
61-233-RR 61-233-RM 61-233-RT
CA62111-158EA 831.12 CAD
CA62111-158 CA62111-160 CA62111-162
Corning® Tris Base Buffer, Powder, Corning
Buffers Electrophoresis Buffers Running Buffers
Tris is commonly used in buffer solutions in biochemical and molecular biological research
This primary amine has an effective pH range of 7.0 - 9.2 and is reported to inhibit a number of different enzymes, limiting its usefulness in protein studies to some extent. This tris base buffer is provided in powder form.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage temperature Ambient
Shipping temperature Ambient