Polyethylene glycol 300 for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: PEG 300

M-Clarity™ quality level = MQ200

8.07484.1000 8.07484.5000
CA8.07484.1000CS 472 CAD
CA8.07484.1000 CA8.07484.5000
Polyethylene glycol 300 for synthesis, Sigma-Aldrich®
Polyethylene glycol 300

8.07484.1000 is offered as an alternative to px1286A-2. different size, different packaging, same chem spec.

Formula: H(OCH₂CH₂)nOH
MW: 300 g/mol
Boiling Pt: >220 °C (1013 hPa) (dec.)
Melting Pt: –15…–10 °C
Density: 1.13 g/cm3 (20 °C)
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR