Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh ASTM) for column chromatography, Millipore®

Supplier: MilliporeSigma

1.07734.2500 1.07734.1000 1.07734.5000 1.07734.1000
CA10199-528EA 276.81 CAD
CA10199-528 EM1.07734.1000 EM1.07734.5000 CA1.07734.1000
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh ASTM) for column chromatography, Millipore®
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
MDL Number: MFCD00011232
CAS Number: 63231-67-4

Order Now

Specification Test Results

pH-value (10 % suspension) 6.5 - 7.5
Fe (Iron) ≤ 0.02 %
Pore volume (N₂-isotherm) 0.74 - 0.84 ml/g
Specific surface area (according to BET; 5-Pt. measurement) 480 - 540 m²/g
Loss on drying (150 °C) ≤ 7.0 %
Particle Size (d10) 75 - 95 µm
Particle Size (d50) 125 - 150 µm
Particle Size (d90) 215 - 245 µm
The specific surface area and the pore volume are related to the raw material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR