Sodium chloride 99.5-100.5% (by argentometric analysis, on dry substance), EMPROVE® EXPERT USP, JP, Ph. Eur., BP, SAFC®

Supplier: EMD MILLIPORE – PCS CA EXCEP

Synonyms: Saline, NaCl, Sea salt, Halite

1.37017.9012 1.37017.9029
CA1.37017.9012EA 1394.87 CAD
CA1.37017.9012 CA1.37017.9029
Sodium chloride 99.5-100.5% (by argentometric analysis, on dry substance), EMPROVE® EXPERT USP, JP, Ph. Eur., BP, SAFC®
Sodium chloride

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Density: 2.17 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Assay (argentometric; calculated on dried substance) 99.5 - 100.5 %
Identity passes test
Appearance of solution passes test
Acidity or alkalinity passes test
Bromide (Br) ≤ 0.005 %
Ferrocyanides passes test
Iodide (I) passes test
Nitrite (NO₂) (UV-absorption 354nm; 1cm; 10%; water) ≤ 0.01
Phosphate (PO₄) ≤ 0.0025 %
Sulfate (SO₄) ≤ 0.0200 %
Heavy metals (as Pb) ≤ 0.0003 %
Al (Aluminium) ≤ 0.00002 %
As (Arsenic) ≤ 0.0001 %
Ba (Barium) passes test
Ca (Calcium) ≤ 0.002 %
Fe (Iron) ≤ 0.0002 %
K (Potassium) ≤ 0.01 %
Magnesium and alkaline-earth metals (as Ca) ≤ 0.0100 %
Residual solvents (ICH Q3C) excluded by production process
Loss on drying (105 °C, 2 h) ≤ 0.5 %
Endotoxins (kinetic turbidimetric test) ≤ 2.5 I.U./g
Microbiological purity (Total aerobic microbial count (TAMC)) ≤ 100.0 CFU/g
Microbiological purity (Total yeast and mould count (TYMC)) ≤ 100.0 CFU/g
DNases (Exo- and endonucleases) (not detectable) passes test
RNases (not detectable) passes test
Proteases (not detectable) passes test
Nicking Activity (not detectable) passes test
Corresponds to Ph Eur,BP,JP,USP


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR