Benzoyl peroxide, (∼25% H₂O) stabilized, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.01641.0100
8.01641.0250
8.01641.1000
8.01641.9026
CA8.01641.0100EA
42
CAD
CA8.01641.0100
CA8.01641.0250
CA8.01641.1000
CA8.01641.9026
Benzoyl peroxide, (∼25% H₂O) stabilized, Sigma-Aldrich®
Benzoyl peroxide
Formula:
C₁₄H₁₀O₄ Melting Pt: 100…105 °C (dec.) Density: 1.33 g/cm3 at 25 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003071 CAS Number: 94-36-0 EINECS: 202-327-6 UN: 3104 ADR: 5.2, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...