Potassium methanolate 30 - 35% in methanol, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Potassium methoxide, Potassium methylate

M-Clarity™ quality level = MQ200

8.14222.0100 8.14222.9025 8.14222.9025
CA8.14222.0100EA 238 CAD
CA8.14222.0100 CA8.14222.9025 EM8.14222.9025
Potassium methanolate 30 - 35% in methanol, Sigma-Aldrich®
Potassium methanolate
Formula: CH₃KO
MW: 70.13 g/mol
Boiling Pt: 93.5 °C (1013 hPa)
Melting Pt: <–20 °C
Density: 0.99 g/cm3 at 20 °C
MDL Number: MFCD00012416
CAS Number: 865-33-8
EINECS: 212-736-1
UN: 2920
ADR: 8.3,II

Order Now

Specification Test Results

Assay (acidimetric) 30-35%
Density (d 20°/4°) 0.985-0.995

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR