Water, Cell Culture grade, tested USP Injection, sterile, Corning®
Supplier: Corning
Synonyms:
Hydrogen oxide
25-055-LB
25-055-LH
25-055-CI
25-055-CV
25-055-CM
25-055-CVC
25-055-LG
CA45000-674EA
772.93
CAD
CA45000-674
CA45001-088
CA95000-092
CA45000-672
CA45001-042
CA89412-362
CA45001-270
Water, Cell Culture grade, tested USP Injection, sterile, Corning®
Water
Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and laboratory reagents.
- Wide array of testing
- High-quality water
- Cell culture media preparation solvent
Cell culture grade water goes through a wide array of testing, which include testing to the chemical and physical properties found in the USP and EP monographs for sterile water for Injection.
Formula:
H₂O MW: 18.02 g/mol Boiling Pt: 100 Melting Pt: 0 Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 Merck Index: 13,10098 |
Specification Test Results
Mycoplasma | Tested negative |
Plating efficiency | Pass |
Oxidizable substances Solution | remains faintly pink |
Total organic carbon (before packaging) USP <643> | < 500 ppb |
Water conductivity (packaged water) USP <645> | Meets USP Requirements |
Water conductivity (before packaging) USP <645> | Meets USP temperature/conductivity requirements |
Endotoxin USP <85> | ≤ 0.025 EU/mL |
Sterility USP <71> | Pass |
Particulate matter USP <788> |
≤ 25 particles/mL that are ≥ 10 μm in size ≤ 3 particles/mL that are ≥ 25 μm in size |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...