E. cloni® Electrocompetent Cells, Biosearch Technologies
Supplier: Lucigen
60052-1 60052-2 60052-3 60052-4 60061-1 60061-2 60080-1 60080-2 60117-1 60117-2 60081-1 60051-1 80026-1
89002-658EA 709.44 CAD
89002-658 89002-660 89002-662 89002-664 89002-666 89002-668 89002-670 89002-672 89002-684 89002-686 95024-012 95024-014 CA89005-074 CA89005-076 89047-690
E. cloni® Electrocompetent Cells, Biosearch Technologies
Competent Cells
Lucigen’s E. cloni competent cells share the most useful genetic elements of standard cloning strains like DH5α™, DH10B™, JM109, TOP10, etc. and directly replace them in cloning protocols. However, E. cloni electrocompetent cells incorporate a unique manufacturing technology that increases transformation efficiency, recombinant yields, and reliability, while decreasing costs. These cells are unrivalled in quality, efficiency, and exceptionally low costs. Lucigen's E. cloni cells provide solutions for a wide range of applications at remarkably economical prices.
Lucigen offers theMore Product Information