Corning® RPMI 1640, Corning
Supplier: Corning
10-040-CM 10-040-LX 10-040-LB 15-040-CV 15-040-CM 15-040-LB 17-105-CV 10-041-CM 17-104-CI 15-041-CV 50-020-PB 50-020-PC 10-043-CV 10-040-CV 10-041-CV 10-104-CV
CA45000-398PK 446.25 CAD
CA45000-398 CA45000-400 CA45000-402 CA45000-404 CA45000-406 CA45000-408 CA45000-410 CA45000-414 CA45000-416 CA45000-418 CA45000-602 CA45000-604 CA45001-110 CA45000-396 CA45000-412 CA71002-878
Corning® RPMI 1640, Corning
Cell Culture Media
Named for the site at which it was developed, the Roswell Park Memorial Institute, RPMI 1640 is a modification of McCoy's 5A containing an increased level of inositol.
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
RPMI 1640 is also used for suspension cultures or monolayer human leukemic cells. Examples of cell types for which this medium is appropriate include mammalian myeloma and hybridoma cells such as mouse hybridomas, human leukocytes, and B and T lymphocytes.