Dichloroacetic acid (DCA) ≥99%, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: DCADCA Deblock

M-Clarity™ quality level = MQ200

DX0657-4000 DX0657-4000
CADX0657-4000CS 15248.33 CAD
CADX0657-4000 EM-DX0657-4000
Dichloroacetic acid (DCA) ≥99%, Sigma-Aldrich®
Dichloroacetic acid (DCA)
Formula: C₂H₂Cl₂O₂
MW: 128.94 g/mol
Boiling Pt: 194 °C (1013 hPa)
Melting Pt: ∼–4 °C
Density: 1.563 g/cm³ (20 °C)
MDL Number: MFCD00004223
CAS Number: 79-43-6
UN: 1764
ADR: 8,II
Merck Index: 13,03075

Order Now

Specification Test Results

Absorbance at 310 nm Report onlyAU
Assay (titration) 99% min
Form Clear liquid
Freezing point 12°C min
Identity (IR-spectrum) Monochloroacetic acid
0.20% max Trichloroacetic acid
1.00% max Water (H2O)
500ppm max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR