Ammonia solution 28.0 - 30.0%, GR ACS, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ300
AX1303-3
AX1303-6
AX1303P-5
CAAX1303-3CS
702
CAD
CAAX1303-3
CAAX1303-6
CAAX1303P-5
Ammonia solution 28.0 - 30.0%, GR ACS, Supelco®
Ammonia solution
Formula:
NH₃ Boiling Pt: 32 °C Melting Pt: –72 °C Density: 0.9 g/ml (25°C) (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00066650 CAS Number: 1336-21-6 EINECS: 215-647-6 UN: 2672 ADR: 8,III |
Specification Test Results
Aluminum (Al) | 0.3ppm max |
Appearance | passes test |
Assay (as NH3) | 28.0-30.0% |
Calcium (Ca) | 0.3ppm max |
Carbon dioxide (CO2) | 0.002% max |
Chloride (Cl) | 0.5ppm max |
Chromium (Cr) | 0.3ppm max |
Copper (Cu) | 0.1ppm max |
Gold (Au) | 0.3ppm max |
Heavy metals (as Pb) | 0.5ppm max |
Iron (Fe) | 0.2ppm max |
Lead (Pb) | 0.2ppm max |
Magnesium (Mg) | 0.3ppm max |
Manganese (Mn) | 0.2ppm max |
Nickel (Ni) | 0.1ppm max |
Nitrate (NO3) | 2ppm max |
Phosphate (PO4) | 2ppm max |
Potassium (K) | 0.3ppm max |
Residue after ignition | 0.002% max |
Sodium (N/A) | 0.3ppm max |
Substances reducing permangaN/Ate | To pass test |
Sulfate (SO4) | 2ppm max |
Tin (Sn) | 0.3ppm max |
Titanium (Ti) | 0.3ppm max |
Zinc (Zn) | 0.3ppm max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...