Paraffin, viscous Reag. Ph. Eur., Supelco®
Supplier: MilliporeSigma
Synonyms:
Wax Paraffin, Paraffin wax
M-Clarity™ quality level = MQ200
1.07160.1000
1.07160.9026
CA1.07160.1000EA
137.39
CAD
CA1.07160.1000
CA1.07160.9026
Paraffin, viscous Reag. Ph. Eur., Supelco®
Paraffin
Boiling Pt:
300…500 °C (1,013 hPa) Density: 0.86 g/cm3 (20 °C) Storage Temperature: Ambient |
CAS Number:
8002-74-2 EINECS: 232-384-2 Merck Index: 13,07263 |
Specification Test Results
Identity (IR-spectrum) | passes test |
Identity (Viscosity) | passes test |
Acidity or alkalinity | passes test |
Relative Density (d 20/20) | 0.827 - 0.890 |
Dynamic viscosity (20 °C) | 110 - 230 mPa∙s |
Aromatic, polycyclic Hydrocarbons | passes test |
Solid Paraffines | passes test |
Readily carbonisable substance | passes test |
Sulfur compounds | passes test |
Corresponds Ph Eur, BP, USP | - |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...