Sodium acetate trihydrate 99,5-101,0 Perchloric acid titration, EMPROVE® ESSENTIAL USP, JP, E262, Ph. Eur., BP, FCC, extra_pure, SAFC®

Supplier: EMD MILLIPORE – PCS CA EXCEP

Synonyms: Acetic acid sodium salt trihydrate

1.06265.9051 1.06265.9525
CA1.06265.9051EA 1688.85 CAD
CA1.06265.9051 CA1.06265.9525
Sodium acetate trihydrate 99,5-101,0 Perchloric acid titration, EMPROVE® ESSENTIAL USP, JP, E262, Ph. Eur., BP, FCC, extra_pure, SAFC®
Sodium acetate trihydrate

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Certifications: Corresponds to Ph. Eur., BP, JP, USP, FCC, E 262
Conforms to the purity criteria on food additives according to the current European Commission Regulation.

Formula: H₃CCOONa·3H₂O
MW: 136.08 g/mol
Boiling Pt: 120…123 °C (1013 hPa)
Melting Pt: 58
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (perchloric acid titration, calculated on dried substance) 99.5 - 101.0 %
Identity passes test
Appearance of solution passes test
In water insoluble matter ≤ 0.03 %
Alkalinity ≤ 0.05 %
Acidity or alkalinity passes test
pH-value (1 %, water) 8.0 - 9.5
pH-value (5 %; water) 7.5 - 9.0
Chloride (Cl) ≤ 0.005 %
Sulfate (SO₄) ≤ 0.0030 %
Heavy metals (as Pb) ≤ 0.0006 %
Al (Aluminium) ≤ 0.00002 %
As (Arsenic) ≤ 0.0002 %
Ca (Calcium) ≤ 0.002 %
Fe (Iron) ≤ 0.0010 %
Hg (Mercury) ≤ 0.0001 %
K (Potassium) ≤ 0.005 %
Mg (Magnesium) ≤ 0.001 %
Pb (Lead) ≤ 0.0002 %
Ca + Mg (calcium and magnesium) passes test
free acetic acid ≤ 0.4 %
Other residual solvents (ICH Q3C) excluded by manufacturing process
formic acid, formate and other oxidizable impurities ≤ 0.1000 %
Substances reducing potassium permanganate (as HCOOH) passes test
Loss on drying (130 °C) 39.0 - 40.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR