Phenol 99.0-100.5%, EMPROVE® ESSENTIAL USP, Ph. Eur., ChP, JP, extra pure, SAFC®

Supplier: EMD MILLIPORE – PCS CA EXCEP
Danger

1.00201.1000 1.00201.9025 1.00201.1000 1.00201.9025
CA1.00201.1000CS 1260.97 CAD
CA1.00201.1000 CA1.00201.9025 EM1.00201.1000 EM1.00201.9025
Phenol 99.0-100.5%, EMPROVE® ESSENTIAL USP, Ph. Eur., ChP, JP, extra pure, SAFC®
Phenol

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Formula: C₆H₅OH
Boiling Pt: 181.7 °C (1013 hPa)
Melting Pt: 40.8 °C
Density: 1.0722 g/cm³ (20 °C)
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1,II
REACH: 01-2119471329-32
Merck Index: 12,07390

Order Now

Specification Test Results

Assay (bromatometric) 99.0 - 100.5 %
Assay (bromatometric, calc. on anhydrous substance) 99.0 - 100.5 %
Identity (IR-spectrum) passes test
Identity (wet chemistry) passes test
Appearance colourless or faintly pink or faintly yellowish crystals or crystalline masses, deliquescent
Appearance of solution (67 g/l, water) clear and not more intensely coloured than reference solution B6
Acidic substances passes test
Solidification temperature ≥ 39.5 °C
Fe (Iron) ≤ 1 ppm
Benzene (HS-GC) ≤ 2 ppm
Methanol (HS-GC) ≤ 3000 ppm
Acetone (HS-GC) ≤ 2 ppm
Anisole ≤ 10 ppm
Cumene (HS-GC) ≤ 70 ppm
Methylisobutylketone (HS-GC) ≤ 2 ppm
Other residual solvents (ICH (Q3C)) excluded by manufacturing process
Residue on evaporation (105 °C, 1 h) ≤ 0.05 %
Residue on evaporation (105 °C) ≤ 0.05 %
Water (according to Karl Fischer) ≤ 0.5 %
Corresponds to Ph. Eur., ChP, JP, USP


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR