Water AquaPur™ USP, purified

Supplier: DECON LAB CA

9301 9305 9355
CA71003-548CS 452 CAD
CA71003-548 CA71003-550 CA71003-552
Water AquaPur™ USP, purified
Water

AquaPur™ Purified Water meets USP and multi-compendial specification, made for applications requiring USP/EP grade Purified Water.



  •  

  •  


AquaPur™ Purified Water has been tested to meet USP/EP specifications and packaged in convenient point-of-use packaging for use in laboratory or production applications requiring purified water.  Made via deionization, reverse osmosis, UV treatment, and ultrafiltration, AquaPur™ meets specification including microbial specification through its 2 year shelf life.



Made via deionization, reverse osmosis, UV treatment, and ultrafiltration, AquaPur meets specification including microbial specification through its 2 yr shelf life.



Packaging:

Packaged in gallon, 5 gallon, and 55 gallon configurations.



Caution:

Not for parenteral use.  Not for injection, wound irrigation or infusion.


Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

USP Water Conductivity USP 645 1.3 umhos/cm At the time of manufacturing
USP Total Organic Carbon USP 643 0.5 ppm
Oxidizable Substances Conforms
Bioburden 100 cfu/mL


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR