Alcohol Burner, WHEATON, DWK Life Sciences
Supplier: DWK Life Sciences
WHEATON®
237070
16299-098PK 665.04 CAD
16299-098
Alcohol Burner, WHEATON, DWK Life Sciences
Heaters Spirit Lamps
The WHEATON Alcohol Burner is manufactured from soda-lime glass and is designed for use with isopropyl or denatured ethyl alcohol
  • The burner's low heat flame is well suited for microscopy purposes
  • Reservoir holds 100 mL of alcohol
  • Glass cap is used to reduce evaporation of alcohol when not in use
Dia.× H: 75×100 mm