Water, Molecular biology grade_meetSpec_USP, sterile, Corning®

Supplier: Corning

Synonyms: Hydrogen oxide

46-000-CM 46-000-CI 46-000-CV
CA45001-044PK 412.43 CAD
CA45001-044 CA95000-094 CA71002-726
Water, Molecular biology grade_meetSpec_USP, sterile, Corning®
Water

Molecular biology grade water is ideal for a wide variety of routine tasks requiring a high degree of sterility Molecular biology grade water is ideal for the preparation of reagents, rinsing glassware and plasticware, and other molecular biology applications where RNase-, DNase-, and Protease-free water is required. No toxic agents, including DEPC, are used in the preparation of Mediatech's molecular biology grade water, eliminating possible interferences with enzymatic reactions.


Certifications: Molecular Biology Grade Water Tested to USP Sterile Purified Water Specifications

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR