Corning® Lymphocyte Separation Medium, Corning
Supplier: Corning
Corning®
25-072-CI 25-072-CV 25-072-CV 25-072-CI
CA45000-728EA 266.93 CAD
CA45000-728 45000-728 CA45000-726 45000-726
Corning® Lymphocyte Separation Medium, Corning
Cell Separation Media
Lymphocyte Separation Medium (LSM) was originally designed for the in vitro isolation of lymphocytes from diluted whole blood
It is a sterile, iso-osmotic polysucrose and diatrizoate solution with low viscosity and a density of 1.077-1.080 g/mL at 20ºC.