Corning® DMEM, Iscove's Modification of DMEM, Corning
Supplier: Corning
10-016-CV 10-016-CM 15-016-CV
CA45000-366PK 330.16 CAD
CA45000-366 CA45000-368 CA45000-370
Corning® DMEM, Iscove's Modification of DMEM, Corning
Cell Culture Media
Iscove's Modification of DMEM (IMDM) is highly enriched synthetic medium optimized for rapidly proliferating cultures of high cell density
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
A modified form of Dulbecco's Modified Eagle Medium (DMEM), Iscove's contains additional amino acids, vitamins, selenium and HEPES buffer, and is formulated with potassium nitrate in place of ferric nitrate. The addition of albumin and transferrin may be required for certain cell types.