Corning® Ham's F-12 Medium, Corning
Supplier: Corning
10-080-CV 10-080-CM 50-040-PB
CA45000-358PK 288.71 CAD
CA45000-358 CA45000-360 CA45000-550
Corning® Ham's F-12 Medium, Corning
Cell Culture Media
Ham's F-12 media was originally formulated for the serum-free culture of CHO cells, and is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids
  • "Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media formulations are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment"
This medium is also suitable for carcinoma cells, rat skeletal myoblasts, Chinese hamster lung cells, and rat, rabbit, and chicken embryos.