O-Rings, CAPFE, Ace Glass Incorporated
Supplier: Ace Glass
7855-805 7855-806 7855-808 7855-810 7855-813 7855-815 7855-816 7855-817 7855-818 7855-819 7855-820 7855-821 7855-822 7855-824 7855-826 7855-827 7855-828 7855-829 7855-830 7855-832 7855-834 7855-838 7855-840 7855-842 7855-844 7855-845 7855-846 7855-847 7855-848 7855-850 7855-861 7855-864 7855-870 7855-871 7855-872 7855-873 7855-874 7855-875 7855-876 7855-877 7855-878 7855-879 7855-880 7855-881 7855-883 7855-884 7855-885 7855-887 7855-889 7855-860 7855-836
89061-080EA 53.95 CAD
89061-080 89061-082 89061-084 89061-086 89061-088 89061-090 89061-092 89061-094 89061-096 89061-098 89061-102 89061-104 89061-106 89061-108 89061-110 89061-112 89061-114 89061-116 89061-118 89061-120 89061-122 89061-124 89061-126 89061-128 89061-130 89061-132 89061-134 89061-136 89061-138 89061-140 89061-142 89061-144 89061-146 89061-148 89061-150 89061-152 89061-154 89061-156 89061-158 89061-160 89061-162 89061-164 89061-166 89061-168 89061-170 89061-172 89061-174 89061-176 89061-178 89084-358 10767-326
O-Rings, CAPFE, Ace Glass Incorporated
Seals Sealing Rings
CAPFE o-rings have a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation
Continuous encapsulation of thick, pure PTFE offers no seams or weak spots to break and/or leak. Will work for a wide variety of applications. Resistant to all chemicals except molten alkali metals, hot fluorine, and certain complex halogenated compounds. The extreme slipperiness of PTFE reduces friction in dynamic applications.
CAPFE ranges from -73°C to +204°C, deterioration vacuum to 10,000 psi. Generally orange in color.
They feature an excellent compression set with excellentMore Product Information