Midget Impinger with Ace-Thred Inlet/Outlet, Ace Glass Incorporated
Supplier: Ace Glass
7533-08 7533-15 7533-23
89059-324EA 310.78 CAD
89059-324 89059-330 89059-336
Midget Impinger with Ace-Thred Inlet/Outlet, Ace Glass Incorporated
Samplers Air Samplers
This piece is used for sampling air volumes at low jet velocity
Bottle joint is reinforced [ST] 24/40 outer. Tube uses #7 "Ace-Safe" removable hose connections for easy connect/disconnect. Nozzle is calibrated to deliver 0.09 to 0.11 CFM at 30.5cm (12 inches) H20 vacuum. Bottle has encircling graduations from 0 to 25mL, or 0 to 30mL, in 5mL divisions. Fabricated from borosilicate glass.
Ordering information: Complete item consists of bottle, tube, connectors and bushings. Items can also be purchased separately.