Springs, Stainless Steel, Ace Glass
Supplier: Ace Glass
8030-02 8030-04 8030-08 8030-12 8030-16 8030-20 8030-24 8030-30
89061-666PK 88.45 CAD
89061-666 89061-668 89061-670 89061-672 89061-674 89061-676 89061-678 89061-680
Springs, Stainless Steel, Ace Glass
Springs
These stainless steel springs can be used to connect interchangeable joints, washing bottles, and other apparatus where glass hooks are provided
The length value refers to coil length.