Visual Scientifics Hooke's Law
Supplier: Wards
PTHOKL PTHOKLSPRING
470135-562EA 48.5 CAD
470135-562 470332-240
Visual Scientifics Hooke's Law
Educational Materials Physics Educational Materials Mechanics Learning Activities
Students investigate and learn the principles of Hooke’s Law with this activity.
  • Lab manual included, with activities and extensions
  • Covers NGSS and common core standards
  • Platform neutral - works with your preferred probeware/video analysis tools
Using technology to investigate Hooke's Law, students will discover the linear relationship between applied mass and spring extension. The included lab manual provides approximately 3 hours of laboratory activities. Instructions are written to explicitly cover Next Generation Science Standards and Common Core concepts.
It's especially applicable to mechanics and physics classes.
Ordering information: Visual Scientifics Base (470148-822) required, but sold separately.
Delivery information: This kit includes 3 springs with different spring constants, aluminum support rod with hook, metal ruler, hanging mass hook with pointer, and 9 to 10 g masses. Recommended accessories: video camera and graphical analysis software.