Corning® Disposable Aseptic Transfer Cap for Disposable Spinner Flask, Corning
Supplier: Corning
3562 3565 3567 3528 3545 3558 3564
89089-908CS 570.25 CAD
89089-908 89089-912 89089-914 89176-434 89176-436 89176-438 89089-910
Corning® Disposable Aseptic Transfer Cap for Disposable Spinner Flask, Corning
Closures Bottle Closures
These disposable Corning® spinner flask aseptic transfer caps will both reduce contamination and enhance the productivity of your cell culture operations when adding or removing medium and cells.
  • Cap is made of high density polyethylene; autoclaving is not recommended
  • The venting port ends in a 25 mm diameter vent filter with a 0.2 µm pore PVDF membrane and a barbed fitting
  • Chemically resistant, heat sealable flexible tubing, ending in a male medical plastic coupler (MPC) connector closed with a female sealing cap
Aseptic transfer caps are sterile (10⁻⁶ SAL), individually double bagged and fit the side arms on 1 L Corning® disposable spinner flasks (Cat. No. 89089-754). Cap is made of high density polyethylene; autoclaving is not recommended. The venting port ends in a 25 mm diameter vent filter with a 0.2 µm pore PVDF membrane and a barbed fitting. The liquid transfer port has an internal end with a rigid polypropylene dip tube angled to reach the bottom of the flask and on the external side a ¹/₈" ID (3.2 mm), ¹/₄" OD chemically resistant, heat sealable flexible tubing, ending in a male medical plastic coupler (MPC) connector closed with a female sealing cap.
Packaging: Aseptic transfer caps are sterile (10⁻⁶ SAL), individually double bagged.