ESK® Sampling Kit, with Tube, Swab and Buffered Peptone Water, Puritan Medical Products
Supplier: Puritan Medical Products
25-83004-PDBPW 25-83004PDBBPW 25-83010-PDBPW 25-83010PDBBPW
89221-718CS 149.28 CAD
89221-718 89221-720 89221-722 89221-724
ESK® Sampling Kit, with Tube, Swab and Buffered Peptone Water, Puritan Medical Products
Swabs Swabs for Sampling
Easy-to-use, reliable test kits are available with Neutralizing Buffer, Buffered Peptone Water, Letheen Broth, or Butterfields Solution, in a 4mL or 10mL fill
These kits are used for the pre-enrichment of injured Salmonella species from food specimens prior to inoculation to a selective enriched medium for isolation and identificaiton.
Pre-filled with 4mL or 10mL Buffered Peptone Water. Shelf life is 18 months. Made in USA.
They come with a small polyester tip attached to a quick-turn, leak-resistant cap to ensure optimum control.
Ordering information: Kits are packaged 50More Product Information