ESK® Sampling Kit, with Tube, Swab and Neutralizing Buffer, Puritan Medical Products
Supplier: Puritan Medical Products
25-83004-PDNB 25-83010-PDBNB 25-83004-PDNB 25-83010-PDNB 25-83004-PDBNB
89221-714CS 158.66 CAD
89221-714 89221-716 89221-710 CA89221-710 89221-712
ESK® Sampling Kit, with Tube, Swab and Neutralizing Buffer, Puritan Medical Products
Swabs Swabs for Sampling
Easy-to-use, reliable test kits are available with Neutralizing Buffer, Buffered Peptone Water, Letheen Broth, or Butterfields Solution, in a 4mL or 10mL fill
Neutralizing buffer is recommended for use in microbiological examination of surfaces for food and dairy products.
Pre-filled with 4mL or 10mL Neutralizing Buffer. Shelf life is 18 months. Made in USA.
They come with a small polyester tip attached to a quick-turn, leak-resistant cap to ensure optimum control.
Ordering information: Kits are packaged 50 individually wrapped tubes per case. Bulk packaging includes 6 boxes of 50 individually wrapped tubes per box.