ESK® Sampling Kit, with Tube, Swab and Butterfield's Solution, Puritan Medical Products
Supplier: Puritan Medical Products
25-83004-PDBS 25-83004-PDBBS 25-83010-PDBS 25-83010-PDBBS
89221-694CS 151.08 CAD
89221-694 89221-696 89221-698 89221-700
ESK® Sampling Kit, with Tube, Swab and Butterfield's Solution, Puritan Medical Products
Swabs Swabs for Sampling
Easy-to-use, reliable test kits are available with Neutralizing Buffer, Buffered Peptone Water, Letheen Broth, or Butterfields Solution, in a 4 ml or 10 ml fill
Butterfield's solution is recommended for the dilution and enumeration of microorganisms in water, food, dairy, and pharmaceutical products.
Pre-filled with 4 ml or 10 ml Butterfield's Solution. Shelf life is 18 months. Made in USA.
They come with a small polyester tip attached to a quick-turn, leak-resistant cap to ensure optimum control.
Ordering information: Kits are packaged 50 individually wrapped tubes per case. Bulk packaging includes 6 boxes of 50 individually wrapped tubes per box.