Puritan® Histobrush® Cytology Collection Devices, Puritan Medical Products
Supplier: Puritan Medical Products
2199 25-2199 2188 25-2188 2199 2188 25-2188 25-2199
CA10805-198CS 6425.61 CAD
CA10805-198 CA10808-158 CA10808-142 CA10808-144 10805-198 10808-142 10808-144 10808-158
Puritan® Histobrush® Cytology Collection Devices, Puritan Medical Products
Swabs Swabs for Clinical Use
Puritan® cytology brushes feature tapered Histobrush® tips made out of soft nylon bristles, attached to a long polystyrene handle
They are used to collect cervical specimens during pap smear tests, as well as buccall cells for DNA lineage.
Certifications: CE marked for medical use. Made in the USA.
Ordering information: Sterile brushes are individually wrapped.