VWR® Applicators with Foam Tip and PP Shaft
Supplier: VWR
1273BKVWR
89176-450CS 922.87 CAD
89176-450
VWR® Applicators with Foam Tip and PP Shaft
Swabs Swabs for Clinical Use
Swabs have thermal edge seals and thermal bonding to the handle help ensure the sponge-like foam will clean up unwanted particles without releasing any into your critical environment.
  • Polypropylene fabrication
  • Safely clean up unwanted particles
Thermal edge seals and thermal bonding help bind particles during cleanup.