PurSwab® Microfiber Tipped Swab, Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
3677 3130 3677 3130
CA89087-856CS 1864.49 CAD
CA89087-856 CA89133-808 89087-856 89133-808
PurSwab® Microfiber Tipped Swab, Polypropylene Handle, Puritan Medical Products
Swabs Swabs for Clinical Use
These microfiber swabs are non-abrasive and lint- and residue-free
Safe to use on and around static-sensitive components. Microfiber tips are soft and non-abrasive, with very low particulates. Made in USA.
Swab 89133-808 is thermally bonded to polypropylene handle, and specifically designed for fiber optics. Swab 89087-856 is ultrasonically welded to a molded polypropylene handle without adhesives.
In addition, the microfiber tip can easily stand up to IPA, acetone, and other solvents commonly used in cleanrooms and other critical environments.
Ordering information: Swabs are packaged 100 swabs per bag, 10 bags per case.