VWR® Upright Freezer Racks for 15 and 50 ml Centrifuge Tubes, Stainless Steel
Supplier: VWR International
89214-720 89214-722 89214-724 89214-726
89214-720EA 579.28 CAD
89214-720 89214-722 89214-724 89214-726
VWR® Upright Freezer Racks for 15 and 50 ml Centrifuge Tubes, Stainless Steel
Racks Cryogenic Box Racks
Durable stainless steel freezer racks facilitate organization and maximize valuable storage space.
  • Racks accommodate 15 ml and 50 ml centrifuge tubes
  • Reorder by simply scanning the QR code with smartphone or tablet
  • Sturdy handles
  • Stainless steel construction
Racks feature sturdy handles and come standard with identification labels for inventory purposes.