VWR® Upright Freezer Racks for 96 Well Microtube Boxes, Stainless Steel
Supplier: VWR International
CP-UFTB-54L CP-UFTB-55L CP-UFTB-64L CP-UFTB-65L
89214-712EA 457.34 CAD
89214-712 89214-714 89214-716 89214-718
VWR® Upright Freezer Racks for 96 Well Microtube Boxes, Stainless Steel
Racks Cryogenic Box Racks
Durable stainless steel freezer racks facilitate organization and maximize valuable storage space.
  • Stainless steel construction
  • Sturdy handle design
  • Easy to reorder
Racks feature sturdy handles. Racks accommodate 96 well microtube boxes and contain locking rods. Reorder by simply scanning the QR code with your smartphone or tablet.