Media Lab Bottle, Non-Graduated, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
219419 219577 219499 219417 219495 219579 219575 W219420 W219500 W219580 219885 219415 W219580
14216-222CS 569.58 CAD
14216-222 14231-278 14231-280 14231-290 16154-715 16158-081 16364-982 16364-984 89200-070 89200-074 89200-076 16152-154 16154-718 66012-903
Media Lab Bottle, Non-Graduated, WHEATON®, DWK Life Sciences
Bottles Cell Culture Bottles
Wheaton media bottles are manufactured from Wheaton 400 borosilicate glass that conforms to USP Type I requirements and have a low alkali content to help prevent changes in pH to maintain the purity of its contents.
  • Hand grips on the 1 L bottle makes for easy handling
The pour lip and sloped shoulders facilitate pouring and cleaning. Pre-attached black phenolic screw caps with liner are attached to each bottle.
Order Now
SPECIFICATIONS
Environmentally Preferable