Contrex® AW, Powder Detergent, Decon Labs
Supplier: Decon Labs
5604 5625 5650
CA89234-830CS 1175 CAD
CA89234-830 CA89234-832 76199-132
Contrex® AW, Powder Detergent, Decon Labs
Cleaning Agents Detergents and Surface Cleansers
Contrex® AW is a concentrated, low-foaming, powdered cleanser formulated for use in all automatic labware washing machines.
  • Eliminates detergent build-up in automatic machines
  • Free-rinsing
  • Completely soluble
  • Biodegradable
Contrex® AW is extremely effective at removing virtually any contaminant from the surface of labware, scientific apparatus, and process equipment.