Distillation Head with Condenser Jacket Side Arm, Ace Glass Incorporated
Supplier: Ace Glass
6608-06 6608-20 9359-04 9359-06
89055-696EA 621.26 CAD
89055-696 89055-698 89074-442 89074-444
Distillation Head with Condenser Jacket Side Arm, Ace Glass Incorporated
Distillation Components Distillation Heads
This distillation head has a condenser jacket on the side arm
The thermometer joint is a [ST] 10/30. Side arm is at a 75° angle. Fabricated from borosilicate glass. Thermometer length value denote the Thermometer Immersion. The Jacket length refers to Condenser Jacket Length.