Reaction Flask, Jacketed, Conical Flange, Ace Glass Incorporated
Supplier: Ace Glass
6475-10 6475-25 6475-15 6475-20 6475-310 6475-315 6475-320 6475-325
89054-882EA 1669.74 CAD
89054-882 89054-888 89054-904 89054-984 10098-954 10098-956 10098-958 10098-960
Reaction Flask, Jacketed, Conical Flange, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
Jacketed cylindrical flask with conical neck opening of 100 mm (4")
Inlet and outlet connections are size 28/15 o-ring ball joints, both sealed tangentially, one at top and one at bottom of jacketed section. Fabricated from borosilicate glass.