Neutrad® Neutral pH Liquid Cleaner
Supplier: Decon Labs
3001 3002
CA89234-804CS 2253 CAD
CA89234-804 CA89234-806 CA89234-808
Neutrad® Neutral pH Liquid Cleaner
Cleaning Agents Detergents and Surface Cleansers
Neutrad® is the perfect safer alternative to old-fashioned caustic soap powders in every manual-scrub cleaning procedure.
  • Phosphate-free and chlorine-free
  • Biodegradable
  • Disposable straight to drain
An instantly soluble liquid detergent concentrate with near neutral-pH, Neutrad® is substrate-safe and skin-kind. The phosphate-free nature of Neutrad®, its inherent rinsability characteristics, and broad-spectrum cleaning capabilities are advantages that make it suitable for all critical laboratory cleaning applications.