Dri-Clean™ Powdered Detergent
Supplier: Decon Labs
2501 2502
CA89234-800EA 1889 CAD
CA89234-800 CA89234-802
Dri-Clean™ Powdered Detergent
Cleaning Agents Detergents and Surface Cleansers
Dri-Clean™ is a powdered detergent for use in cleaning all labware and equipment.
  • Completely free-rinsing and residue-free
  • User-friendly bottle provides easy dispensing and eliminates caking
Use Dri-Clean™ for soak, manual, and ultrasonic cleaning applications. It removes protein, dried blood, and most organic and inorganic contaminants from glassware, plasticware, ceramics, and metal surfaces. Packaged in a wide-mouth plastic container with a convenient handle. This packaging format makes it easier to handle and prevents the absorption of water and
More Product Information