Dri-Contrad® Powder Detergent
Supplier: Decon Labs
2005 2002 2004 2007
CA89234-792CS 1305 CAD
CA89234-792 CA89234-788 CA89234-790 CA89234-794
Dri-Contrad® Powder Detergent
Cleaning Agents Detergents and Surface Cleansers
Dri-Contrad® is a non-foaming powder cleaner designed for use exclusively in automatic labware washing machines.
  • Chlorinated for contamination control
  • Non-corrosive to machine components
  • Effective against a wide range of contaminants
  • Rinsable
Dri-Contrad® is freely soluble and will not cake in the machine. Totally rinsable, manufacturer tested, and proven machine-kind, it will not corrode pumps, seals, or liners.