Sodium chloride, Molecular Biology Grade, Tested to USP Specifications
Supplier: IBI Scientific
IB07070
IB07071
IB07072
IB07073
89140-854EA
64.78
CAD
89140-854
89140-856
89140-858
89140-860
Sodium chloride, Molecular Biology Grade, Tested to USP Specifications
Sodium chloride
IBI's Sodium Chloride is an ionic compound with the molecular formula NaCl
Store at room temperature and protect from moisture.
Sodium Chloride is the salt found most in the extracellular fluid of many multicellular organisms. It is also used as the main component in saline solution.
Formula:
NaCl Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 |
Specification Test Results
Appearance | White Crystals |
Assay (on dried basis) | 90.0 - 100.5% |
Bromides | Max. 0.010% |
Phosphates | Max. 0.0025% |
Heavy Metals (as Pb) | Max. 5ppm |
Aresenic (As) | Max. 1 mcg/g |
Iron (Fe) | Max. 2 mcg/g |
Sulfate (SO4) | Max. 0.020% |
Loss on Drying (@1056C / 2hrs) | Max. 0.5% |
Magnesium / Alkaline Earth Metals (as Ca) | Max. 0.01% |
Bioburden | Max. 40CFU/g |
Endotoxin | Max. 0.025EU/ml |
Acidity / Alkalinity | Pass |
Appearance of Solution (in H2O) | Pass |
Ferrocyanides | Pass |
Identification Positive for Na & Cl | Pass |
Identification Positive for Iodides | Pass |
Identification Positive for Barium | Pass |
Identification Positive for Nitrites | Pass |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...