Conflikt® Ready-To-Use Disinfectant
Supplier: Decon Labs
Conflikt®
4102 4104 4101 4103 4104 4101 4102
CA89233-180CS 874.52 CAD
89233-180 CA89233-180 89219-350 89233-182 CA89233-178 89233-178 CA89233-182
Conflikt® Ready-To-Use Disinfectant
Cleaning Agents Disinfectants
Conflikt ® Ready-to-use disinfectant spray cleans and disinfects hard surfaces that may harbor viruses, bacteria and fungi.
Pre-diluted, stable solution of dual quaternary ammonium active ingredients is effective against HIV-1, E-coli and other pathogenic organisms. Conforms to OSHA's Bloodborne Pathogens Standard. Conflikt is effective against a wide range of organisms including TB. EPA registered.