LB Agar Lennox, IBI Scientific
Supplier: IBI Scientific
IB49120 IB49121 IB49122
89126-178EA 216.79 CAD
89126-178 89126-180 89126-182
LB Agar Lennox, IBI Scientific
Media for Microbiology Dehydrated Media for Microbiology
IBI growth media products start with raw materials sourced only from North America and European suppliers, this insures the highest lot-to-lot quality and consistency
IBI's LB Agar Lennox is composed of 10.0gm Casein Digest Peptone, 5.0gm Sodium Chloride, 5.0gm Yeast Extract, and 15.0gm Agar with a pH (@25°C) of 7.0 ± 0.2. To prepare, mix 35.0gm of the medium in 1L of purified water until evenly dispersed. Heat with repeated stirring and boil for 1 minute to dissolve completely. Distribute and autoclave at 121.0°C for 15 minutes.
Store at room temperature, keep tightly sealed, and protect from moisture and light.
IBI's LB Agar Lennox is used in molecular genetic studies. It is a nutritionally rich media for the growth and maintenance of pure cultures of recombinant strains of E. coli. The inclusion of casein peptone and yeast extract supply essential growth factors, such as nitrogen, carbon, sulfur, minerals, and vitamins. Sodium chloride also provides essential electrolytes. Agar is used as a solidification agent.