Contrad® 70 Liquid Detergent
Supplier: Decon Labs
1003 1003 1004 1002 1005
42000-000CS 1110.16 CAD
42000-000 CA42000-000 CA89233-154 CA89233-152 CA89233-156
Contrad® 70 Liquid Detergent
Cleaning Agents Detergents and Surface Cleansers
Contrad® 70 is an emulsion of anionic and non-ionic surfactants in an alkaline aqueous base.
  • Phosphate-free, chlorine-free, and biodegradable
  • Free-rinsing and disposable straight to the drain
  • Cleans glass, plastic, ceramic, and ferrous metals
Contrad® 70 works by soaking, scrubbing, or in ultrasonic cleaning baths. It removes virtually any contaminant from every surface, even silicones, dried proteins, carbon, most resins, waxes, and oils. The formulation is totally rinsable, eliminating detergent build-up that interferes with analysis, tissue culture, and other critical procedures. It is also extremely effective when used as an ambient soak solution, with no need to scrub. Its activity can be accelerated by heating, agitation, and ultrasonics, or by increasing concentrations. Used solutions may be disposed straight to drain providing they have not been used to remove contaminants that are in themselves pollutants.