Ace-Safe Tubing Connector, Polypropylene, Ace Glass Incorporated
Supplier: Ace Glass
5853-03 5853-06 5853-09 5853-10 5853-12 5853-15 5853-18 5853-19 5853-20 5853-21 5853-23 5853-26 5853-31 5853-33 5853-35 5853-37
89052-934EA 39.37 CAD
89052-934 89052-936 89052-940 89052-942 89052-944 89052-946 89052-948 89052-950 89052-952 89052-954 89052-956 89052-958 89052-960 89052-962 89052-964 89052-966
Ace-Safe Tubing Connector, Polypropylene, Ace Glass Incorporated
Fittings and Connectors
These polypropylene tubing connectors are used with nylon bushings to make an easy and safe connect/disconnect of coolant or steam tubing to jacketed reaction flasks, condensers, or any vessel utilizing the Ace-Thred as the connection port
Connector is fitted with a silicone o-ring. Bushing compresses connector and o-ring for leak-tight seal. Once tubing is attached to serrated end of connector, bushing will not come off. Polypropylene connectors have a maximum temperature of 110°C.
Ordering information: Available complete with bushing, connector, and o-ring, or as polypropyleneMore Product Information