Flask, Volumetric, Class A, Ace Glass Incorporated
Supplier: Ace Glass
7124-20 7124-15
89053-114EA 363.33 CAD
89053-114 89057-576
Flask, Volumetric, Class A, Ace Glass Incorporated
Flasks Volumetric Flasks
A specially designed, standardized volumetric flask, with stopper
All capacities have a standard taper 12/18 outer joint with an opening of approximately 10mm. The cylindrical body affords better mixing and drainage, and ease of withdrawal using pipets. The wide base offers greater stability. The upper surface of the base is ground to allow pencil markings. Class “A” tolerances. Fabricated from borosilicate glass.